Assay Details
| Gene Symbol: ATOH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009857.2 | XP_009008105.1 | 2 | 1012 | 66 | |
| XM_009009858.2 | XP_009008106.1 | 4 | 969 | 66 | ||
| XM_017979073.1 | XP_017834562.1 | 4 | 1019 | 66 | ||
| XM_017979074.1 | XP_017834563.1 | 3 | 1427 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009009857.2, XM_009009858.2, XM_017979073.1, XM_017979074.1 transcripts.
Set Membership: |
|

ATOH7
Genomic Map