Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994332.2 | XP_008992580.1 | 9 | 2368 | 65 | |
| XM_008994336.2 | XP_008992584.1 | 9 | 2377 | 65 | ||
More Information
Set Membership: |
|

ETV7
Genomic Map