Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270916.1 | NP_001257845.1 | 2 - 3 | 217 | 127 | |
| XM_008987855.2 | XP_008986103.1 | 3 - 4 | 404 | 127 | ||
| XM_008987857.2 | XP_008986105.1 | 3 - 4 | 1112 | 127 | ||
More Information
Set Membership: |
|

ZFP30
Genomic Map