Assay Details
| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735275.3 | XP_003735323.2 | 3 - 4 | 334 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP1LC3C| Gene Symbol: MAP1LC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735275.3 | XP_003735323.2 | 3 - 4 | 334 | 108 | |
Set Membership: |
|