Assay Details
| Gene Symbol: WNT8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744194.3 | XP_002744240.2 | 4 - 5 | 570 | 63 | |
| XM_008985444.2 | XP_008983692.1 | 4 - 5 | 570 | 63 | ||
More Information
Set Membership: |
|

WNT8A
Genomic Map