Assay Details
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747909.4 | XP_002747955.2 | 39 - 40 | 5783 | 65 | |
XM_017972280.1 | XP_017827769.1 | 38 - 39 | 5727 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
