Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733869.3 | XP_003733917.1 | 1 - 2 | 972 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS37B| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733869.3 | XP_003733917.1 | 1 - 2 | 972 | 78 | |