Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733869.3 | XP_003733917.1 | 3 - 4 | 1234 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS37B| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733869.3 | XP_003733917.1 | 3 - 4 | 1234 | 91 | |