Assay Details
| Gene Symbol: RAB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762968.4 | XP_002763014.2 | 3 - 4 | 278 | 85 | |
| XM_017968772.1 | XP_017824261.1 | 3 - 4 | 278 | 85 | ||
| XM_017968773.1 | XP_017824262.1 | 3 - 4 | 278 | 85 | ||
More Information
Set Membership: |
|

RAB41
Genomic Map