Assay Details
Gene Symbol: RAB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762968.4 | XP_002763014.2 | 5 - 6 | 497 | 84 | |
XM_017968772.1 | XP_017824261.1 | 5 - 6 | 497 | 84 | ||
XM_017968773.1 | XP_017824262.1 | 5 - 6 | 497 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
