Assay Details
| Gene Symbol: CATSPERB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734064.3 | XP_003734112.3 | 14 - 15 | 1291 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CATSPERB| Gene Symbol: CATSPERB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734064.3 | XP_003734112.3 | 14 - 15 | 1291 | 62 | |
Set Membership: |
|