Assay Details
| Gene Symbol: MARCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963349.1 | XP_017818838.1 | 5 - 6 | 2415 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCH8| Gene Symbol: MARCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963349.1 | XP_017818838.1 | 5 - 6 | 2415 | 63 | |
Set Membership: |
|