Assay Details
| Gene Symbol: TAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748441.3 | XP_002748487.1 | 2 - 3 | 471 | 60 | |
| XM_008997391.2 | XP_008995639.1 | 2 - 3 | 468 | 60 | ||
| XM_008997392.2 | XP_008995640.1 | 2 - 3 | 469 | 60 | ||
| XM_008997393.2 | XP_008995641.1 | 2 - 3 | 438 | 60 | ||
More Information
Set Membership: |
|

TAC4
Genomic Map