Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760808.3 | XP_002760854.2 | 3 - 4 | 646 | 57 | |
| XM_008985670.2 | XP_008983918.1 | 3 - 4 | 646 | 57 | ||
| XM_008985671.2 | XP_008983919.1 | 3 - 4 | 1615 | 57 | ||
| XM_008985674.2 | XP_008983922.1 | 3 - 4 | 892 | 57 | ||
More Information
Set Membership: |
|

TRIM17
Genomic Map