Assay Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994322.2 | XP_008992570.1 | 2 - 3 | 249 | 65 | |
| XM_017971564.1 | XP_017827053.1 | 2 - 3 | 322 | 65 | ||
More Information
Set Membership: |
|

CLPSL2
Genomic Map