Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734917.3 | XP_003734965.2 | 1 - 2 | 1210 | 88 | |
| XM_008982449.2 | XP_008980697.1 | 1 - 2 | 1213 | 88 | ||
| XM_008982451.2 | XP_008980699.1 | 1 - 2 | 1213 | 88 | ||
More Information
Set Membership: |
|

GTPBP8
Genomic Map