Assay Details
| Gene Symbol: LOC100389993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994741.2 | XP_008992989.1 | 1 - 2 | 970 | 105 | |
| XM_017971245.1 | XP_017826734.1 | 1 - 2 | 994 | 105 | ||
More Information
Set Membership: |
|

LOC100389993
Genomic Map