Assay Details
Gene Symbol: LOC100404101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017975211.1 | XP_017830700.1 | 2 - 3 | 2064 | 71 | |
XM_017975214.1 | XP_017830703.1 | 2 - 3 | 2061 | 71 | ||
XM_017975217.1 | XP_017830706.1 | 2 - 3 | 2062 | 71 | ||
XM_017975220.1 | XP_017830709.1 | 2 - 3 | 282 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
