Assay Details
| Gene Symbol: GLYATL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746608.3 | XP_002746654.1 | 6 - 7 | 721 | 74 | |
| XM_017971148.1 | XP_017826637.1 | 6 - 7 | 710 | 74 | ||
| XM_017971149.1 | XP_017826638.1 | 5 - 6 | 614 | 74 | ||
More Information
Set Membership: |
|

GLYATL3
Genomic Map