Assay Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759193.3 | XP_002759239.1 | 2 - 3 | 182 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY6D| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759193.3 | XP_002759239.1 | 2 - 3 | 182 | 76 | |
Set Membership: |
|