Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008989963.2 | XP_008988211.1 | 7 - 8 | 1210 | 71 | |
| XM_008989964.2 | XP_008988212.1 | 7 - 8 | 1207 | 71 | ||
| XM_008989965.2 | XP_008988213.1 | 7 - 8 | 1210 | 71 | ||
| XM_008989966.2 | XP_008988214.1 | 7 - 8 | 1210 | 71 | ||
More Information
Set Membership: |
|

MAP7D3
Genomic Map