Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007499.2 | XP_009005747.1 | 1 - 2 | 350 | 91 | |
| XM_009007501.2 | XP_009005749.1 | 1 - 2 | 350 | 91 | ||
| XM_017977830.1 | XP_017833319.1 | 1 - 2 | 683 | 91 | ||
| XM_017977831.1 | XP_017833320.1 | 1 - 2 | 687 | 91 | ||
More Information
Set Membership: |
|

ART5
Genomic Map