Assay Details
| Gene Symbol: SERPINB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757294.3 | XP_002757340.1 | 7 - 8 | 802 | 72 | |
| XM_002757295.3 | XP_002757341.1 | 5 - 6 | 690 | 72 | ||
| XM_008979935.2 | XP_008978183.1 | 7 - 8 | 832 | 72 | ||
More Information
Set Membership: |
|

SERPINB7
Genomic Map