Assay Details
| Gene Symbol: CLDN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759582.2 | XP_002759628.1 | 3 - 4 | 498 | 63 | |
| XM_008983776.2 | XP_008982024.1 | 3 - 4 | 694 | 63 | ||
More Information
Set Membership: |
|

CLDN18
Genomic Map