Assay Details
| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747892.3 | XP_002747938.1 | 1 | 120 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST3A1| Gene Symbol: HS3ST3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747892.3 | XP_002747938.1 | 1 | 120 | 86 | |
Set Membership: |
|