Assay Details
| Gene Symbol: Kcnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172976.1 | NP_001166447.1 | 2 - 3 | 750 | 86 | |
| GenBank mRNA | AY075096.1 | - | 2 - 3 | 750 | 86 | |
More Information
Set Membership: |
|

Kcnk1
Genomic Map