Assay Details
| Gene Symbol: Fshr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172949.1 | NP_001166420.1 | 8 - 9 | 768 | 67 | |
| XM_005005015.1 | XP_005005072.1 | 7 - 8 | 591 | 67 | ||
| GenBank mRNA | AY082514.1 | - | 8 - 9 | 768 | 67 | |
More Information
Set Membership: |
|

Fshr
Genomic Map