Assay Details
| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172940.1 | NP_001166411.1 | 2 - 3 | 939 | 106 | |
| XM_013145545.1 | XP_013000999.1 | 2 - 3 | 972 | 106 | ||
| GenBank mRNA | D11043.1 | - | 2 - 3 | 939 | 106 | |
More Information
Set Membership: |
|

Cyp1a1
Genomic Map