Assay Details
| Gene Symbol: Htr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172962.2 | NP_001166433.1 | 3 | 100 | ||
| GenBank mRNA | EF423870.1 | - | 3 | 100 | ||
| U69635.1 | - | 1 | 147 | 100 | ||
More Information
Set Membership: |
|

Htr2b
Genomic Map