Assay Details
Gene Symbol: Zp3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173027.1 | NP_001166498.1 | 3 - 4 | 346 | 99 | |
GenBank mRNA | U75654.1 | - | 3 - 4 | 346 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
