Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173027.1 | NP_001166498.1 | 4 - 5 | 440 | 87 | |
| GenBank mRNA | U75654.1 | - | 4 - 5 | 440 | 87 | |
More Information
Set Membership: |
|

Zp3r
Genomic Map