Assay Details
| Gene Symbol: Ubc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173065.1 | NP_001166536.1 | 253 | 103 | ||
| GenBank mRNA | D83208.1 | - | 253 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubc| Gene Symbol: Ubc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173065.1 | NP_001166536.1 | 253 | 103 | ||
| GenBank mRNA | D83208.1 | - | 253 | 103 | ||
Set Membership: |
|