Assay Details
| Gene Symbol: Kcnj8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173123.1 | NP_001166594.1 | 2 - 3 | 457 | 91 | |
| GenBank mRNA | AF183918.1 | - | 2 - 3 | 457 | 91 | |
More Information
Set Membership: |
|

Kcnj8
Genomic Map