Assay Details
| Gene Symbol: Hcar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172752.1 | NP_001166223.1 | 1 | 742 | 74 | |
| GenBank mRNA | AM746460.1 | - | 1 | 742 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcar2| Gene Symbol: Hcar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172752.1 | NP_001166223.1 | 1 | 742 | 74 | |
| GenBank mRNA | AM746460.1 | - | 1 | 742 | 74 | |
Set Membership: |
|