Assay Details
| Gene Symbol: Fth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172847.1 | NP_001166318.1 | 2 - 3 | 394 | 62 | |
| GenBank mRNA | EB366819.1 | - | 2 - 3 | 402 | 62 | |
| EB371193.1 | - | 2 - 3 | 327 | 62 | ||
More Information
Set Membership: |
|

Fth1
Genomic Map