Assay Details
Gene Symbol: cnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038855.3 | NP_001033944.1 | 3 - 4 | 646 | 59 | |
NM_001259363.2 | NP_001246292.1 | 2 - 3 | 347 | 59 | ||
NM_001259364.2 | NP_001246293.1 | 2 - 3 | 347 | 59 | ||
NM_001259365.2 | NP_001246294.1 | 3 - 4 | 424 | 59 | ||
NM_001259366.2 | NP_001246295.1 | 3 - 4 | 335 | 59 | ||
NM_057736.6 | NP_477084.1 | 2 - 3 | 347 | 59 | ||
NM_165990.3 | NP_725297.1 | 3 - 4 | 646 | 59 | ||
GenBank mRNA | BT010053.1 | - | 2 - 3 | 317 | 59 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
