Assay Details
| Gene Symbol: Rcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274033.2 | NP_001260962.1 | 3 - 4 | 1124 | 66 | |
| NM_001274034.2 | NP_001260963.1 | 3 - 4 | 1491 | 66 | ||
| NM_001274035.2 | NP_001260964.1 | 3 - 4 | 1124 | 66 | ||
| NM_001274036.2 | NP_001260965.1 | 3 - 4 | 1491 | 66 | ||
| NM_001299475.1 | NP_001286404.1 | 3 - 4 | 1491 | 66 | ||
| NM_137083.4 | NP_610927.3 | 3 - 4 | 1491 | 66 | ||
| NM_166031.3 | NP_725357.2 | 3 - 4 | 1124 | 66 | ||
| NM_166032.3 | NP_725358.1 | 3 - 4 | 961 | 66 | ||
| GenBank mRNA | AY069502.1 | - | 3 - 4 | 1466 | 66 | |
| BT100232.1 | - | 3 - 4 | 1090 | 66 | ||
More Information
Set Membership: |
|

Rcd1
Genomic Map