Assay Details
| Gene Symbol: DCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014585.2 | NP_001014585.1 | 3 - 4 | 719 | 71 | |
| NM_001259847.2 | NP_001246776.1 | 2 - 3 | 1694 | 71 | ||
| NM_140548.5 | NP_648805.2 | 2 - 3 | 1694 | 71 | ||
| NM_168634.5 | NP_730074.2 | 2 - 3 | 879 | 71 | ||
| GenBank mRNA | BT004829.1 | - | 2 - 3 | 860 | 71 | |
| BT016065.1 | - | 2 - 3 | 1670 | 71 | ||
More Information
Set Membership: |
|

DCP2
Genomic Map