Assay Details
| Gene Symbol: eIF4H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298402.1 | NP_001285331.1 | 4 - 5 | 1112 | 54 | |
| NM_080220.5 | NP_524959.1 | 4 - 5 | 837 | 54 | ||
| NM_167522.5 | NP_727982.1 | 4 - 5 | 834 | 54 | ||
| NM_206764.3 | NP_996487.1 | 4 - 5 | 936 | 54 | ||
| GenBank mRNA | BT088437.1 | - | 4 - 5 | 923 | 54 | |
More Information
Set Membership: |
|

eIF4H1
Genomic Map