Assay Details
Gene Symbol: Ntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135291.3 | NP_609135.1 | 5 - 6 | 1136 | 57 | |
NM_164765.4 | NP_723303.1 | 5 - 6 | 927 | 57 | ||
GenBank mRNA | BT133486.1 | - | 5 - 6 | 924 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
