Assay Details
Gene Symbol: Ntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135291.3 | NP_609135.1 | 7 - 8 | 1415 | 82 | |
NM_164765.4 | NP_723303.1 | 7 - 8 | 1206 | 82 | ||
GenBank mRNA | BT133486.1 | - | 7 - 8 | 1203 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
