Assay Details
| Gene Symbol: Agpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169441.2 | NP_001162912.1 | 3 - 4 | 1027 | 52 | |
| NM_001298826.1 | NP_001285755.1 | 2 - 3 | 273 | 52 | ||
| NM_135387.4 | NP_609231.2 | 2 - 3 | 579 | 52 | ||
| NM_164823.3 | NP_723398.1 | 1 - 2 | 297 | 52 | ||
| GenBank mRNA | AJ238252.1 | - | 2 - 3 | 534 | 52 | |
| BT001684.1 | - | 2 - 3 | 575 | 52 | ||
| BT044180.1 | - | 2 - 3 | 563 | 52 | ||
| BT072915.1 | - | 3 - 4 | 702 | 52 | ||
More Information
Set Membership: |
|

Agpat2
Genomic Map