Assay Details
Gene Symbol: UQCR-C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275026.1 | NP_001261955.1 | 1 - 2 | 171 | 111 | |
NM_140648.2 | NP_648905.1 | 1 - 2 | 320 | 111 | ||
GenBank mRNA | AY089228.1 | - | 1 - 2 | 304 | 111 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
