Assay Details
| Gene Symbol: UQCR-C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275026.1 | NP_001261955.1 | 2 - 3 | 307 | 58 | |
| NM_140648.2 | NP_648905.1 | 2 - 3 | 456 | 58 | ||
| GenBank mRNA | AY089228.1 | - | 2 - 3 | 440 | 58 | |
More Information
Set Membership: |
|

UQCR-C2
Genomic Map