Assay Details
Gene Symbol: UQCR-C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275026.1 | NP_001261955.1 | 3 - 4 | 615 | 126 | |
NM_140648.2 | NP_648905.1 | 3 - 4 | 764 | 126 | ||
GenBank mRNA | AY089228.1 | - | 3 - 4 | 748 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
