Assay Details
| Gene Symbol: kug | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031967.2 | NP_001027138.2 | 18 - 19 | 13706 | 64 | |
| NM_001300197.1 | NP_001287126.1 | 18 - 19 | 13825 | 64 | ||
| NM_140914.3 | NP_649171.3 | 18 - 19 | 13706 | 64 | ||
More Information
Set Membership: |
|

kug
Genomic Map