Assay Details
| Gene Symbol: Dyrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038817.1 | NP_001033906.1 | 7 - 8 | 1154 | 75 | |
| NM_001038818.1 | NP_001033907.1 | 7 - 8 | 1327 | 75 | ||
| NM_001273526.1 | NP_001260455.1 | 7 - 8 | 1400 | 75 | ||
| NM_001273527.1 | NP_001260456.1 | 3 - 4 | 747 | 75 | ||
| NM_078840.5 | NP_523564.1 | 6 - 7 | 1322 | 75 | ||
| NM_205988.3 | NP_995710.1 | 7 - 8 | 1418 | 75 | ||
| NM_205989.3 | NP_995711.1 | 7 - 8 | 1414 | 75 | ||
| GenBank mRNA | AF168467.1 | - | 7 - 8 | 1389 | 75 | |
| BT010088.1 | - | 6 - 7 | 1322 | 75 | ||
| BT133412.1 | - | 3 - 4 | 693 | 75 | ||
More Information
Set Membership: |
|

Dyrk2
Genomic Map