Assay Details
| Gene Symbol: mRpS18B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078882.3 | NP_523606.1 | 1 - 2 | 391 | 59 | |
| NM_165321.2 | NP_724248.1 | 1 - 2 | 391 | 59 | ||
| GenBank mRNA | AY075438.1 | - | 2 - 3 | 359 | 59 | |
More Information
Set Membership: |
|

mRpS18B
Genomic Map