Assay Details
| Gene Symbol: Dsor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298115.1 | NP_001285044.1 | 2 - 3 | 912 | 64 | |
| NM_078543.4 | NP_511098.1 | 2 - 3 | 1208 | 64 | ||
| GenBank mRNA | AY058692.1 | - | 2 - 3 | 755 | 64 | |
More Information
Set Membership: |
|

Dsor1
Genomic Map