Assay Details
| Gene Symbol: Pp2B-14D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258786.3 | NP_001245715.1 | 1 | 3051 | 108 | |
| NM_001258787.2 | NP_001245716.1 | 1 | 3051 | 108 | ||
| NM_078649.4 | NP_523373.2 | 1 | 3051 | 108 | ||
| GenBank mRNA | BT010083.1 | - | 1 | 3058 | 108 | |
More Information
Set Membership: |
|

Pp2B-14D
Genomic Map